Product Details

SNP ID
rs9914271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30905607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAACTAAATAAAATTAAATTTT[A/G]TACGTTTATAAGAAATGTTTCAGCT
Phenotype
MIM: 609534 MIM: 616422
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAD5 PubMed Links

Gene Details

Gene
ATAD5
Gene Name
ATPase family, AAA domain containing 5
There are no transcripts associated with this gene.

Gene
TEFM
Gene Name
transcription elongation factor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024683.3 Intron NP_078959.3
XM_006722084.1 Intron XP_006722147.1

View Full Product Details