Product Details
- SNP ID
-
rs9333029
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:46930490 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCCGTCCTGGCTTCACACTATCCC[A/G]ATCCTTGGACAGAGCCGTCTTCCTG
- Phenotype
-
MIM: 601310
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYP4A11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9333032,rs9333033] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYP4A11
- Gene Name
- cytochrome P450 family 4 subfamily A member 11
View Full Product Details