Product Details

SNP ID
rs10465605
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:115982694 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTGTTTATCAGTCTTCTCAAGC[C/T]CAGACTTCTCCTGATTCCATTCATA
Phenotype
MIM: 608275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928977 PubMed Links

Gene Details

Gene
LOC101928977
Gene Name
uncharacterized LOC101928977
There are no transcripts associated with this gene.

Gene
SLC22A15
Gene Name
solute carrier family 22 member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018420.2 Intron NP_060890.2
XM_005271004.3 Intron XP_005271061.1

View Full Product Details