Product Details
- SNP ID
-
rs10137162
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:69778005 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACTCTTGCGAATAGAGGCCTTTCA[C/G]ACAACTAATAACCTCAGTTGTCCAA
- Phenotype
-
MIM: 182396
MIM: 600914
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC10A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116123638] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC10A1
- Gene Name
- solute carrier family 10 member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003049.3 |
|
Intron |
|
|
NP_003040.1 |
- Gene
- SRSF5
- Gene Name
- serine and arginine rich splicing factor 5
There are no transcripts associated with this gene.
View Full Product Details