Product Details

SNP ID
rs6127785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56542042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTAGGTAAGTCAATCCCAAAAC[C/T]AGGGAGGTGACGCGGAGCAACGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM209A PubMed Links

Gene Details

Gene
FAM209A
Gene Name
family with sequence similarity 209 member A
There are no transcripts associated with this gene.

Gene
FAM209B
Gene Name
family with sequence similarity 209 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013646.3 Intron NP_001013668.2
XM_017027849.1 Intron XP_016883338.1

View Full Product Details