Product Details

SNP ID
rs6520260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47389108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTACAGGTGTGAACCACCACACCC[A/G]GCCGGAAACAATATTTTCAATTACA
Phenotype
MIM: 300024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORA11C PubMed Links

Gene Details

Gene
SNORA11C
Gene Name
small nucleolar RNA, H/ACA box 11C
There are no transcripts associated with this gene.

Gene
ZNF157
Gene Name
zinc finger protein 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003446.3 Intron NP_003437.2

View Full Product Details