Product Details
- SNP ID
-
hCV29674968
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35618338 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATACTGGGAGGTCAAGGAGGAGTC[G/T]AGGAAAGGGGACACAGTCTTCAAAA
- Phenotype
-
MIM: 604205
MIM: 603038
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE1
PubMed Links
Gene Details
- Gene
- CPNE1
- Gene Name
- copine 1
There are no transcripts associated with this gene.
- Gene
- SPAG4
- Gene Name
- sperm associated antigen 4
There are no transcripts associated with this gene.
View Full Product Details