Product Details

SNP ID
rs6088193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33362089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGAGGTGGGTGGAAGAGGAA[A/G]CGGAGCAAAAAAATCTGAAGAGTAA
Phenotype
MIM: 608200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK5RAP1 PubMed Links

Gene Details

Gene
CDK5RAP1
Gene Name
CDK5 regulatory subunit associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278167.1 Intron NP_001265096.1
NM_001278168.1 Intron NP_001265097.1
NM_001278169.1 Intron NP_001265098.1
NM_016082.4 Intron NP_057166.4
NM_016408.3 Intron NP_057492.2
XM_011528856.2 Intron XP_011527158.1
XM_011528857.2 Intron XP_011527159.1
XM_011528858.1 Intron XP_011527160.1
XM_011528859.2 Intron XP_011527161.1
XM_011528860.1 Intron XP_011527162.1
XM_017027876.1 Intron XP_016883365.1
XM_017027877.1 Intron XP_016883366.1

View Full Product Details