Product Details

SNP ID
rs6057813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33285122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGAAGCTTTATAGGGAAAGTGA[A/C]AAATCTTGGCAGGGTTTTAAAGGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BPIFB1 PubMed Links

Gene Details

Gene
BPIFB1
Gene Name
BPI fold containing family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033197.2 Intron NP_149974.2

View Full Product Details