Product Details

SNP ID
rs6053145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5136808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTCCTTCCAATTCTTGTTTTA[A/G]GTTCAGGGGATACATGTGCAGGTTT
Phenotype
MIM: 603549
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDS2 PubMed Links

Gene Details

Gene
CDS2
Gene Name
CDP-diacylglycerol synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003818.3 Intron NP_003809.1
XM_006723660.1 Intron XP_006723723.1
XM_011529390.1 Intron XP_011527692.1

View Full Product Details