Product Details

SNP ID
rs10424390
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57125169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGGTGAAGCCAGGCTTTCCATG[C/T]AGTTGTGTGGTTTTTAGTGAGTTTC
Phenotype
MIM: 609546
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
USP29 PubMed Links

Gene Details

Gene
USP29
Gene Name
ubiquitin specific peptidase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020903.2 Intron NP_065954.1
Gene
ZIM3
Gene Name
zinc finger imprinted 3
There are no transcripts associated with this gene.

View Full Product Details