Product Details

SNP ID
rs10235312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64512838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAAAGCTCTAATTAAAAATAAG[C/T]GCTTAAATCAATTTTTTTAAAATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF680 PubMed Links

Gene Details

Gene
ZNF680
Gene Name
zinc finger protein 680
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130022.1 Intron NP_001123494.1
NM_178558.4 Intron NP_848653.2
XM_006715959.2 Intron XP_006716022.1
XM_017012126.1 Intron XP_016867615.1
XM_017012127.1 Intron XP_016867616.1
XM_017012128.1 Intron XP_016867617.1
XM_017012129.1 Intron XP_016867618.1

View Full Product Details