Product Details

SNP ID
rs9824133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121773618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCAAATATTCCTGTTCCTATC[C/T]TGGATATGACCCTGTTCCACTAAAA
Phenotype
MIM: 609237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCB1 PubMed Links

Gene Details

Gene
IQCB1
Gene Name
IQ motif containing B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023570.3 Intron NP_001018864.2
NM_001023571.3 Intron NP_001018865.2
NM_001319107.1 Intron NP_001306036.1
XM_005247911.3 Intron XP_005247968.1
XM_005247912.2 Intron XP_005247969.1
XM_011513335.2 Intron XP_011511637.1
XM_017007537.1 Intron XP_016863026.1
XM_017007538.1 Intron XP_016863027.1
XM_017007539.1 Intron XP_016863028.1

View Full Product Details