Product Details

SNP ID
rs7876431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111693130 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTGGCTAATGGCTAATTCCCTA[C/T]GTGAATGCTCTATTTATTATACAGT
Phenotype
MIM: 300776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG13 PubMed Links

Gene Details

Gene
ALG13
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039210.4 Intron NP_001034299.3
NM_001099922.2 Intron NP_001093392.1
NM_001168385.2 Intron NP_001161857.1
NM_001257230.1 Intron NP_001244159.1
NM_001257231.1 Intron NP_001244160.1
NM_001257234.1 Intron NP_001244163.1
NM_001257235.2 Intron NP_001244164.1
NM_001257237.1 Intron NP_001244166.1
NM_001257239.2 Intron NP_001244168.1
NM_001257240.2 Intron NP_001244169.1
NM_001257241.2 Intron NP_001244170.1
NM_001324290.1 Intron NP_001311219.1
NM_001324291.1 Intron NP_001311220.1
NM_001324292.1 Intron NP_001311221.1
NM_001324293.1 Intron NP_001311222.1
NM_001324294.1 Intron NP_001311223.1
NM_018466.5 Intron NP_060936.1
XM_006724693.2 Intron XP_006724756.1
XM_006724697.3 Intron XP_006724760.2
XM_006724698.3 Intron XP_006724761.2
XM_011531028.2 Intron XP_011529330.1
XM_011531030.1 Intron XP_011529332.1
XM_011531031.1 Intron XP_011529333.1
XM_011531032.1 Intron XP_011529334.1
XM_011531033.2 Intron XP_011529335.1
XM_011531034.2 Intron XP_011529336.1
XM_011531037.1 Intron XP_011529339.1
XM_017029846.1 Intron XP_016885335.1
XM_017029847.1 Intron XP_016885336.1
XM_017029848.1 Intron XP_016885337.1
XM_017029849.1 Intron XP_016885338.1
XM_017029850.1 Intron XP_016885339.1
XM_017029851.1 Intron XP_016885340.1
XM_017029852.1 Intron XP_016885341.1
XM_017029853.1 Intron XP_016885342.1
XM_017029854.1 Intron XP_016885343.1
XM_017029855.1 Intron XP_016885344.1

View Full Product Details