Product Details

SNP ID
rs9814749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10301588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAATAACAGCAGCTGTGGATGA[C/T]GATGTCAACACAGGATACTGTTTTC
Phenotype
MIM: 605353 MIM: 600152
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHRL PubMed Links

Gene Details

Gene
GHRL
Gene Name
ghrelin/obestatin prepropeptide
There are no transcripts associated with this gene.

Gene
GHRLOS
Gene Name
ghrelin opposite strand/antisense RNA
There are no transcripts associated with this gene.

Gene
SEC13
Gene Name
SEC13 homolog, nuclear pore and COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136026.2 Intron NP_001129498.1
NM_001136232.2 Intron NP_001129704.1
NM_001278946.1 Intron NP_001265875.1
NM_030673.3 Intron NP_109598.2
NM_183352.2 Intron NP_899195.1
XM_005265379.2 Intron XP_005265436.1
XM_017007019.1 Intron XP_016862508.1
XM_017007020.1 Intron XP_016862509.1
XM_017007021.1 Intron XP_016862510.1

View Full Product Details