Product Details
- SNP ID
-
rs10129218
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:67371158 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCCAGGCACGGTGGCTCACACTTA[C/T]AATTCCAGCAATTTCGGATGCTGAG
- Phenotype
-
MIM: 603907
MIM: 603930
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF2S1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10140203,rs72719111] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF2S1
- Gene Name
- eukaryotic translation initiation factor 2 subunit alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004094.4 |
|
Intron |
|
|
NP_004085.1 |
- Gene
- GPHN
- Gene Name
- gephyrin
View Full Product Details