Product Details

SNP ID
rs10494870
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:205507061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCTACACTGTAGGGCAGAGCCCG[A/G]CTTGACTCTTCCCGGTTCCTACCCT
Phenotype
MIM: 169190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK18 PubMed Links

Gene Details

Gene
CDK18
Gene Name
cyclin dependent kinase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002596.3 Intron NP_002587.2
NM_212502.2 Intron NP_997667.1
NM_212503.2 Intron NP_997668.1
XM_011509602.2 Intron XP_011507904.1
XM_017001423.1 Intron XP_016856912.1

View Full Product Details