Product Details
- SNP ID
-
rs5994554
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:32361496 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAATAATTAAATCTTTATGCAGA[C/G]ATTAATAAATATGCAGGCTAACAAA
- Phenotype
-
MIM: 605970
MIM: 605971
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RFPL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149059829] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RFPL3
- Gene Name
- ret finger protein like 3
- Gene
- RFPL3S
- Gene Name
- RFPL3 antisense
There are no transcripts associated with this gene.
- Gene
- SLC5A4
- Gene Name
- solute carrier family 5 member 4
There are no transcripts associated with this gene.
View Full Product Details