Product Details

SNP ID
rs6065349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41402591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTTGCAGTAAGGCTTTCAAGA[C/T]ACAGGAATTTTTATAGCATTTGTAT
Phenotype
MIM: 616114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHD6 PubMed Links

Gene Details

Gene
CHD6
Gene Name
chromodomain helicase DNA binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032221.4 10328 UTR 3 NP_115597.3

View Full Product Details