Product Details
- SNP ID
-
rs6070741
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38602442 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGGCCCAGGGGCTCAAGATTCAG[C/G]AGTCTCTGGGTCAAGTTTCTGGGTC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116568379] are located under a probe and SNP(s) [rs116351315] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGAP40
- Gene Name
- Rho GTPase activating protein 40
View Full Product Details