Product Details

SNP ID
rs9875555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33804710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACACAGGAGATTTAAGCCCACA[A/C]AGCTATCTTTATGGCAGAACTGGAC
Phenotype
MIM: 608074
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDCD6IP PubMed Links

Gene Details

Gene
PDCD6IP
Gene Name
programmed cell death 6 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162429.2 Intron NP_001155901.1
NM_001256192.1 Intron NP_001243121.1
NM_013374.5 Intron NP_037506.2
XM_011533252.1 Intron XP_011531554.1
XM_011533253.1 Intron XP_011531555.1
XM_017005488.1 Intron XP_016860977.1

View Full Product Details