Product Details

SNP ID
rs9655764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99157723 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAAATTTCTTGTCAGGTAAGAA[A/C]GCAAGCCATTGTGATTCTTGATCTT
Phenotype
MIM: 614107
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KPNA7 PubMed Links

Gene Details

Gene
KPNA7
Gene Name
karyopherin subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516215.2 Intron XP_011514517.1
XM_017012208.1 Intron XP_016867697.1
XM_017012209.1 Intron XP_016867698.1
XM_017012210.1 Intron XP_016867699.1
XM_017012211.1 Intron XP_016867700.1
XM_017012212.1 Intron XP_016867701.1
XM_017012213.1 Intron XP_016867702.1
XM_017012214.1 Intron XP_016867703.1
XM_017012215.1 Intron XP_016867704.1
XM_017012216.1 Intron XP_016867705.1

View Full Product Details