Product Details

SNP ID
rs10117756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:95807932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCCTGAATCCTTAGGAAAGGTG[C/G]ACAGCTGGTTTCTGCCTTCAAACAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00476 PubMed Links

Gene Details

Gene
LINC00476
Gene Name
long intergenic non-protein coding RNA 476
There are no transcripts associated with this gene.

View Full Product Details