Product Details
- SNP ID
-
rs8179144
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108622746 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGAGAAGCGGGGAGAGCCATGGGA[G/A]ATGTTAGAGGGACCCAGAGAAGATG
- Phenotype
-
MIM: 600738
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SELPLG
PubMed Links
Gene Details
- Gene
- SELPLG
- Gene Name
- selectin P ligand
View Full Product Details