Product Details

SNP ID
rs9646856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218453840 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTAAATGCCCAAGGTTTGTCCTA[C/T]AGCTCAAGTACTTAATTTCATACTT
Phenotype
MIM: 193040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP37 PubMed Links

Gene Details

Gene
USP37
Gene Name
ubiquitin specific peptidase 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020935.2 4407 UTR 3 NP_065986.2
XM_005246720.2 4407 UTR 3 XP_005246777.1
XM_005246721.3 4407 UTR 3 XP_005246778.1
XM_005246722.3 4407 UTR 3 XP_005246779.1
XM_005246724.1 4407 UTR 3 XP_005246781.1
XM_011511538.2 4407 UTR 3 XP_011509840.1
XM_011511541.2 4407 Intron XP_011509843.1
XM_017004577.1 4407 UTR 3 XP_016860066.1
Gene
VIL1
Gene Name
villin 1
There are no transcripts associated with this gene.

View Full Product Details