Product Details

SNP ID
rs9677975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127482705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGCCTCAGCAATCAGGGCACACA[C/T]GGGCCAGTGGGTTACAAAATATATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IWS1 PubMed Links

Gene Details

Gene
IWS1
Gene Name
IWS1, SUPT6H interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017969.2 Intron NP_060439.2
XM_005263707.3 Intron XP_005263764.1
XM_006712626.3 Intron XP_006712689.1
XM_011511450.1 Intron XP_011509752.1
XM_011511452.1 Intron XP_011509754.1
XM_017004467.1 Intron XP_016859956.1
XM_017004468.1 Intron XP_016859957.1

View Full Product Details