Product Details

SNP ID
rs10197681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19912152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACTCCACTGGCACGTTACAC[C/T]AACAGTGTTAATCTATCATTGCTTT
Phenotype
MIM: 613602
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR35 PubMed Links

Gene Details

Gene
WDR35
Gene Name
WD repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006657.1 3752 UTR 3 NP_001006658.1
NM_020779.3 3752 UTR 3 NP_065830.2
XM_011533007.2 3752 UTR 3 XP_011531309.1

View Full Product Details