Product Details

SNP ID
rs9859484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:96818670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTCACAAACTCTGATTGAATA[C/T]TGTTCTTATATTTATAGAAATGGAG
Phenotype
MIM: 600066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHA6 PubMed Links

Gene Details

Gene
EPHA6
Gene Name
EPH receptor A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080448.2 Intron NP_001073917.2
NM_001278300.1 Intron NP_001265229.1
NM_001278301.1 Intron NP_001265230.1
NM_173655.3 Intron NP_775926.1
XM_006713592.3 Intron XP_006713655.1
XM_011512705.2 Intron XP_011511007.1
XM_011512706.2 Intron XP_011511008.1
XM_011512707.2 Intron XP_011511009.1
XM_017006210.1 Intron XP_016861699.1
XM_017006211.1 Intron XP_016861700.1
XM_017006212.1 Intron XP_016861701.1
XM_017006213.1 Intron XP_016861702.1
XM_017006214.1 Intron XP_016861703.1
XM_017006215.1 Intron XP_016861704.1
XM_017006216.1 Intron XP_016861705.1
XM_017006217.1 Intron XP_016861706.1
XM_017006218.1 Intron XP_016861707.1
XM_017006219.1 Intron XP_016861708.1
XM_017006220.1 Intron XP_016861709.1
XM_017006221.1 Intron XP_016861710.1
XM_017006222.1 Intron XP_016861711.1
XM_017006223.1 Intron XP_016861712.1

View Full Product Details