Product Details

SNP ID
rs9879736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139518046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGGGAGCAAATGTTTGTTCTG[C/T]CACTTTCTAGTTGTGCAATTTGGAC
Phenotype
MIM: 180260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507291 PubMed Links

Gene Details

Gene
LOC100507291
Gene Name
uncharacterized LOC100507291
There are no transcripts associated with this gene.

Gene
RBP1
Gene Name
retinol binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130992.1 Intron NP_001124464.1
NM_001130993.1 Intron NP_001124465.1
NM_002899.3 Intron NP_002890.2

View Full Product Details