Product Details
- SNP ID
-
rs7258844
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14439483 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCATGGGAATGGGTACACTGTGAGC[C/T]CATTTTCCAGGCACACACCTGCAGG
- Phenotype
-
MIM: 601032
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PKN1
PubMed Links
Gene Details
- Gene
- PKN1
- Gene Name
- protein kinase N1
View Full Product Details