Product Details

SNP ID
rs5908762
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143714172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGTCTCACACCCAACCCCCTTT[C/T]GCTAGGAACCAGGCACATGCCCAAA
Phenotype
MIM: 300665
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SPANXN2 PubMed Links

Gene Details

Gene
SPANXN2
Gene Name
SPANX family member N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009615.2 Intron NP_001009615.1

View Full Product Details