Product Details

SNP ID
rs10477484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112861443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGTTCTGAGGCGGTGGGTCCTC[A/C]GAAAGGAAGGGGCTTGCTGTGGTGC
Phenotype
MIM: 182175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRP19 PubMed Links

Gene Details

Gene
SRP19
Gene Name
signal recognition particle 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204193.1 Intron NP_001191122.1
NM_001204194.1 Intron NP_001191123.1
NM_001204196.1 Intron NP_001191125.1
NM_001204199.1 Intron NP_001191128.1
NM_003135.2 Intron NP_003126.1

View Full Product Details