Product Details

SNP ID
rs9991702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128875351 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAGAACTGTAGAGAACATATCA[A/C]ATTTGTGCAGAGCACAGATTCACTG
Phenotype
MIM: 610514 MIM: 611399
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JADE1 PubMed Links

Gene Details

Gene
JADE1
Gene Name
jade family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287437.1 3806 Intron NP_001274366.1
NM_001287439.1 3806 Intron NP_001274368.1
NM_001287440.1 3806 Intron NP_001274369.1
NM_001287441.1 3806 Intron NP_001274370.1
NM_001287442.1 3806 Intron NP_001274371.1
NM_001287443.1 3806 Intron NP_001274372.1
NM_024900.4 3806 Intron NP_079176.2
NM_199320.3 3806 Intron NP_955352.1
XM_005263232.1 3806 Intron XP_005263289.1
XM_017008626.1 3806 Intron XP_016864115.1
XM_017008627.1 3806 Intron XP_016864116.1
Gene
SCLT1
Gene Name
sodium channel and clathrin linker 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300897.1 3806 Intron NP_001287826.1
NM_001300898.1 3806 Intron NP_001287827.1
NM_144643.3 3806 Intron NP_653244.2
XM_017007716.1 3806 Intron XP_016863205.1
XM_017007717.1 3806 UTR 3 XP_016863206.1
XM_017007718.1 3806 Intron XP_016863207.1
XM_017007719.1 3806 Intron XP_016863208.1
XM_017007720.1 3806 Intron XP_016863209.1
XM_017007721.1 3806 Intron XP_016863210.1

View Full Product Details