Product Details

SNP ID
rs6099708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38447364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTTTACAGACCAGTTGGCTGA[A/G]GTTTAAATGGAGGACGCTCCCAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNHG11 PubMed Links

Gene Details

Gene
SNHG11
Gene Name
small nucleolar RNA host gene 11
There are no transcripts associated with this gene.

Gene
SNORA60
Gene Name
small nucleolar RNA, H/ACA box 60
There are no transcripts associated with this gene.

Gene
SNORA71E
Gene Name
small nucleolar RNA, H/ACA box 71E
There are no transcripts associated with this gene.

View Full Product Details