Product Details

SNP ID
rs10407367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47274964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGGCGGCGCCGAGAGCCCCGG[A/C]GGCGCGGGGCTGAGCGAGGGCCCGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC9 PubMed Links

Gene Details

Gene
CCDC9
Gene Name
coiled-coil domain containing 9
There are no transcripts associated with this gene.

Gene
INAFM1
Gene Name
InaF motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178511.5 80 Silent Mutation GGA,GGC G15G NP_848606.3

View Full Product Details