Product Details

SNP ID
rs6018145
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46902182 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGTGGACAATTTTTCTAACACC[C/T]TGGGACCCGATTTTGGAAATGTCTT
Phenotype
MIM: 601654
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EYA2 PubMed Links

Gene Details

Gene
EYA2
Gene Name
EYA transcriptional coactivator and phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005244.4 Intron NP_005235.3
NM_172110.3 Intron NP_742108.2
XM_005260327.2 Intron XP_005260384.1
XM_017027721.1 Intron XP_016883210.1

View Full Product Details