Product Details
- SNP ID
-
hCV29745357
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70029260 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACACAATTGACAGTCACAAACAA[C/T]GGTAAAATTGTCTTTTTATATCATA
- Phenotype
-
MIM: 142702
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HTN3
PubMed Links
Gene Details
- Gene
- HTN3
- Gene Name
- histatin 3
There are no transcripts associated with this gene.
View Full Product Details