Product Details

SNP ID
rs9596220
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49996055 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTATCGACTGAGTAACTGTTAG[C/T]GGTGGATAATTTAAAGCCAATTTGA
Phenotype
MIM: 605766 MIM: 605661
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DLEU2 PubMed Links

Gene Details

Gene
DLEU2
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MIR3613
Gene Name
microRNA 3613
There are no transcripts associated with this gene.

Gene
TRIM13
Gene Name
tripartite motif containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007278.2 Intron NP_001007279.1
NM_005798.4 Intron NP_005789.2
NM_052811.3 Intron NP_434698.1
NM_213590.2 Intron NP_998755.1

View Full Product Details