Product Details

SNP ID
hCV29752084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108563688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTAAAACGTTAACGTAGGAAGG[C/T]GTTTTCACCTTTCCTGTGATTTTTA
Phenotype
MIM: 602681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXO3 PubMed Links

Gene Details

Gene
FOXO3
Gene Name
forkhead box O3
There are no transcripts associated with this gene.

View Full Product Details