Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130058.1 | Intron | NP_001123530.1 | |||
NM_001320283.1 | Intron | NP_001307212.1 | |||
NM_001320285.1 | Intron | NP_001307214.1 | |||
NM_001320287.1 | Intron | NP_001307216.1 | |||
NM_152697.4 | Intron | NP_689910.2 | |||
XM_006710445.3 | Intron | XP_006710508.1 | |||
XM_011540986.2 | Intron | XP_011539288.1 | |||
XM_011540987.2 | Intron | XP_011539289.1 | |||
XM_017000607.1 | Intron | XP_016856096.1 | |||
XM_017000608.1 | Intron | XP_016856097.1 | |||
XM_017000609.1 | Intron | XP_016856098.1 | |||
XM_017000610.1 | Intron | XP_016856099.1 | |||
XM_017000611.1 | Intron | XP_016856100.1 |