Product Details

SNP ID
rs6698208
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:50066153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCAGTTGATCTACCCTACCAC[C/T]CTTACATTCCTTCTAGAAATAAGAG
Phenotype
MIM: 168360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELAVL4 PubMed Links

Gene Details

Gene
ELAVL4
Gene Name
ELAV like neuron-specific RNA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144774.2 Intron NP_001138246.1
NM_001144775.2 Intron NP_001138247.2
NM_001144776.2 Intron NP_001138248.1
NM_001144777.2 Intron NP_001138249.1
NM_001294348.1 Intron NP_001281277.1
NM_001324208.1 Intron NP_001311137.1
NM_001324209.1 Intron NP_001311138.1
NM_001324212.1 Intron NP_001311141.1
NM_001324213.1 Intron NP_001311142.1
NM_001324214.1 Intron NP_001311143.1
NM_001324215.1 Intron NP_001311144.1
NM_001324216.1 Intron NP_001311145.1
NM_001324217.1 Intron NP_001311146.1
NM_021952.4 Intron NP_068771.2
XM_006710411.3 Intron XP_006710474.1
XM_011540889.2 Intron XP_011539191.1
XM_011540890.2 Intron XP_011539192.1
XM_011540893.2 Intron XP_011539195.1
XM_011540894.2 Intron XP_011539196.1
XM_011540895.2 Intron XP_011539197.1
XM_017000538.1 Intron XP_016856027.1
XM_017000539.1 Intron XP_016856028.1
XM_017000540.1 Intron XP_016856029.1
XM_017000541.1 Intron XP_016856030.1
XM_017000542.1 Intron XP_016856031.1

View Full Product Details