Product Details
- SNP ID
-
rs6701046
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:116755112 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCCCCAGTGGAGACTGTGGTCCA[A/G]TGCGGGAATGGGATCCACACATCTG
- Phenotype
-
MIM: 186990
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD2
PubMed Links
Gene Details
- Gene
- CD2
- Gene Name
- CD2 molecule
There are no transcripts associated with this gene.
View Full Product Details