Product Details

SNP ID
rs6030881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43459930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATTCAAGATAGAAATGATATGA[C/T]TAATGCTTTAAAGTAAACTCCTTTT
Phenotype
MIM: 601944
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF6 PubMed Links

Gene Details

Gene
SRSF6
Gene Name
serine and arginine rich splicing factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006275.5 Intron NP_006266.2

View Full Product Details