Product Details

SNP ID
rs10125557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132414410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGGATACATGCACACGTTTATA[A/G]CTGTGCAGATAGAGATGAATGCGAA
Phenotype
MIM: 600777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP77 PubMed Links

Gene Details

Gene
CFAP77
Gene Name
cilia and flagella associated protein 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282957.1 Intron NP_001269886.1
NM_207417.2 Intron NP_997300.1
XM_011518670.2 Intron XP_011516972.1
XM_011518671.1 Intron XP_011516973.1
XM_017014711.1 Intron XP_016870200.1
Gene
TTF1
Gene Name
transcription termination factor 1
There are no transcripts associated with this gene.

View Full Product Details