Product Details

SNP ID
rs10075465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75675150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGCTCAAAGGAACACCTCAGA[C/G]ATGAGCACTGAGTCATTAAAGTCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKDD1B PubMed Links

Gene Details

Gene
ANKDD1B
Gene Name
ankyrin repeat and death domain containing 1B
There are no transcripts associated with this gene.

Gene
POC5
Gene Name
POC5 centriolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099271.1 Intron NP_001092741.1
NM_152408.2 Intron NP_689621.2
XM_005248436.1 Intron XP_005248493.1
XM_011543158.2 Intron XP_011541460.1
XM_011543159.1 Intron XP_011541461.1
XM_011543160.1 Intron XP_011541462.1
XM_017009037.1 Intron XP_016864526.1
XM_017009038.1 Intron XP_016864527.1
XM_017009039.1 Intron XP_016864528.1
XM_017009040.1 Intron XP_016864529.1

View Full Product Details