Product Details

SNP ID
rs6622898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84061743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATACTACCAAAAACCATTAACT[A/G]GAAAAACACAATGCAAAATCATTAT
Phenotype
MIM: 300303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 4808 UTR 3 NP_055311.1
XM_011530917.2 4808 Intron XP_011529219.1
XM_011530918.2 4808 Intron XP_011529220.1
XM_011530919.2 4808 Intron XP_011529221.1
XM_011530920.2 4808 Intron XP_011529222.1
XM_017029423.1 4808 UTR 3 XP_016884912.1
XM_017029424.1 4808 Intron XP_016884913.1
XM_017029425.1 4808 Intron XP_016884914.1
XM_017029426.1 4808 Intron XP_016884915.1

View Full Product Details