Product Details

SNP ID
rs6035521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20025695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTAGTATGGGATTCCTTTCTAC[C/T]TACAATACCTCGCCCACTGGCCAGA
Phenotype
MIM: 610952 MIM: 610833
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRNKL1 PubMed Links

Gene Details

Gene
CRNKL1
Gene Name
crooked neck pre-mRNA splicing factor 1
There are no transcripts associated with this gene.

Gene
NAA20
Gene Name
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016100.4 204 Silent Mutation CTA,TTA L33L NP_057184.1
NM_181527.3 204 Silent Mutation CTA,TTA L21L NP_852668.1
NM_181528.3 204 Silent Mutation CTA,TTA L33L NP_852669.1

View Full Product Details