Product Details

SNP ID
rs10036780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150667140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAGAATAAAAAAAAATCAGTGT[C/G]CTTTTTAGACAGCTATTTAGGATTC
Phenotype
MIM: 610735 MIM: 608155
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYOZ3 PubMed Links

Gene Details

Gene
MYOZ3
Gene Name
myozenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122853.2 Intron NP_001116325.1
NM_133371.4 Intron NP_588612.2
Gene
SYNPO
Gene Name
synaptopodin
There are no transcripts associated with this gene.

View Full Product Details