Product Details

SNP ID
rs9834486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113213342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGTCACCTAGTTCTTTGTTGT[C/T]CAGAACTCGGTACACATTGACCATT
Phenotype
MIM: 608708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOC PubMed Links

Gene Details

Gene
BOC
Gene Name
BOC cell adhesion associated, oncogene regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301861.1 Intron NP_001288790.1
NM_001301867.1 Intron NP_001288796.1
NM_033254.3 Intron NP_150279.1
XM_005247891.2 Intron XP_005247948.1
XM_005247892.2 Intron XP_005247949.1
XM_005247897.2 Intron XP_005247954.1
XM_011513305.2 Intron XP_011511607.1
XM_017007441.1 Intron XP_016862930.1
XM_017007442.1 Intron XP_016862931.1
XM_017007443.1 Intron XP_016862932.1
XM_017007444.1 Intron XP_016862933.1
XM_017007445.1 Intron XP_016862934.1
XM_017007446.1 Intron XP_016862935.1
XM_017007447.1 Intron XP_016862936.1
XM_017007448.1 Intron XP_016862937.1
XM_017007449.1 Intron XP_016862938.1
XM_017007450.1 Intron XP_016862939.1
XM_017007451.1 Intron XP_016862940.1
XM_017007452.1 Intron XP_016862941.1
XM_017007453.1 Intron XP_016862942.1
XM_017007454.1 Intron XP_016862943.1
XM_017007455.1 Intron XP_016862944.1

View Full Product Details