Product Details

SNP ID
rs4721745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19113889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATAACAATTACCTTCTCAGCAGT[C/G]ATGGCCAGCTCCATCCTGGTGTACC
Phenotype
MIM: 601622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TWIST1 PubMed Links

Gene Details

Gene
TWIST1
Gene Name
twist family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000474.3 784 Intron NP_000465.1
XM_011515496.1 784 UTR 3 XP_011513798.1

View Full Product Details